Order Entry
Czech Republic
ContactUsLinkComponent
Potassium chloride ≥99.0%, crystals FCC
Potassium chloride ≥99.0%, crystals FCC
Supplier:  Spectrum Chemical
New Product
Certificates
Potassium chloride ≥99.0%, crystals FCC
Supplier:  Spectrum Chemical
New Product

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (KCl; Dried Basis):
    99.0 % min.
  • Acidity / Alkalinity:
    To Pass Test
  • Heavy Metals (As Pb):
    max. 5 mg/kg
  • Iodide or Bromide:
    To Pass Test
  • Loss On Drying:
    max. 1.0 %

Specification Test Results

Frequently Bought Together