Phenolphthalein 2% in ethanol indicator, Fluka™
Catalog # 34605-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Test of sensitivity:complying
- Change range (pH 7.2-10.0):complying
Specification Test Results
About this item
:
:318.33 g/mol
Boiling Pt:80 °C (1.013 hPa)
Density:0.900 g/cm3 (20 °C)
Flash Pt:24 °C
Storage Temperature:Ambient
EINECS:201-004-7
UN:1170
ADR:3,III
:GHS02
:GHS08
:GHS07
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913