Order Entry
Czech Republic
ContactUsLinkComponent
Orthoboric acid 40 g/l in aqueous solution with indicator, VOLUSOL®
Orthoboric acid 40 g/l in aqueous solution with indicator, VOLUSOL®
Catalog # 95097.5000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Orthoboric acid 40 g/l in aqueous solution with indicator, VOLUSOL®
Catalog # 95097.5000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic jerrican
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay (W/V):
    3.9 - 4.2 %
  • Appearance:
    Clear red liquid
  • Sensitivity:
    Passes test
  • Density (20/4):
    1.000 - 1.020

Specification Test Results

About this item

 :
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337