Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
VWR®, Diazinon 100 µg/ml in methanol, pesticide reference standard
VWR®, Diazinon 100 µg/ml in methanol, pesticide reference standard
VWR®, Diazinon 100 µg/ml in methanol, pesticide reference standard
123972C
:
VWR Chemicals
:
VWR®, Diazinon 100 µg/ml in methanol, pesticide reference standard
123972C
:
VWR Chemicals
:
Pack type:
Glass ampoule
Pk:
1 mL
CAS No.:
333-41-5
Description:
Diazinon 100 µg/ml in methanol
Formula:
C₁₂H₂₁N₂O₃PS
Description:
Diazinon