VWR®, Chlorpyrifos, pesticide reference standard, 100 µg/ml in methanol
Catalog # 123692U
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 mL
- CAS No.:2921-88-2
- Description:Chlorpyrifos 100 µg/ml in methanol
- Formula:C₉H₁₁Cl₃NO₃PS
- Description:Chlorpyrifos