Order Entry
Czech Republic
ContactUsLinkComponent
Calcon® (mordant black 17) indicator for complexometry (Al, Fe, Zr), Fluka™
Irritant
Calcon® (mordant black 17) indicator for complexometry (Al, Fe, Zr), Fluka™
Catalog # 45550-25G
CAS Number:  
Irritant
Calcon® (mordant black 17) indicator for complexometry (Al, Fe, Zr), Fluka™
Catalog # 45550-25G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Indicator solution:
    complying
  • Suitability for Complexometry:
    complying

Specification Test Results

About this item

 :
 :416.38 g/mol
EINECS:219-810-2
 :GHS07
Storage Temperature:Ambient
CAS Number:2538-85-4
Formula:C₂₀H₁₃N₂NaO₅S
MDL Number:MFCD00004069