Order Entry
Czech Republic
ContactUsLinkComponent
Bromothymol blue 0.04% in aqueous solution pH-indicator
Bromothymol blue 0.04% in aqueous solution pH-indicator
Catalog # REGEBRTH05
Supplier:  REAGECON
CAS Number:  
Bromothymol blue 0.04% in aqueous solution pH-indicator
Catalog # REGEBRTH05
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :624.39 g/mol
Storage Temperature:Ambient
EINECS:200-971-2
CAS Number:76-59-5
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872