Methyl red ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:493-52-7
:
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425
Merck Index:14,6119
Melting Pt:179…182 °C
Density:1.31 g/cm³ (20 °C)
:269.3 g/mol
Storage Temperature:Ambient
EINECS:207-776-1
Related Information
Frequently Bought Together