4-Methoxystyrene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:637-69-4
:
Formula:C₉H₁₀O
MDL Number:MFCD00008619
:134.18 g/mol
Boiling Pt:221 °C (1013 hPa)
Melting Pt:3.3 °C
Density:1.002 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:211-298-9
Frequently Bought Together