

1,3-Diaminocyclohexane cis- and trans-mixture 95+%
Catalog # ACRO370240100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥95.0 % (sum of cis and trans)
Specification Test Results
About this item
:
:114.19 g/mol
EINECS:222-194-8
UN:2922
:GHS07
:GHS05
CAS Number:3385-21-5
Formula:C₆H₁₄N₂
MDL Number:MFCD00059563