
3,5-Di-tert-butylpyrocatechol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1020-31-1
:
Formula:C₁₄H₂₂O₂
MDL Number:MFCD00008819
Boiling Pt:310 °C (1013 hPa)
Melting Pt:97…100 °C
:222.33 g/mol
EINECS:213-816-9
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:97 to 100 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together