
Congo red indicator
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:573-58-0
:
Formula:C₃₂H₂₂N₆Na₂O₆S₂
MDL Number:MFCD00004028
:696.67 g/mol
Merck Index:14,2498
Melting Pt:>360 °C
EINECS:209-358-4
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together