ContactUsLinkComponent
Styrene-[D₈] 98+ atom% D stabilised for NMR spectroscopy
Flammable
Irritant
Health
Styrene-[D₈] 98+ atom% D stabilised for NMR spectroscopy
Certificates
undefined
Flammable
Irritant
Health
Styrene-[D₈] 98+ atom% D stabilised for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (Coulometric)
  • Stabilizer:
    0.5 % 4-tert-Butylcatechol
  • D-Enrichment:
    ≥98.0 %