Zadání objednávky
Czech Republic
ContactUsLinkComponent
FLUOR DE LYS®-succinyl, desuccinylase substrate 5 mM ≥95% (by HPLC)
Irritant
FLUOR DE LYS®-succinyl, desuccinylase substrate 5 mM ≥95% (by HPLC)
Catalog # ENZOBMLKI5900050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Irritant
FLUOR DE LYS®-succinyl, desuccinylase substrate 5 mM ≥95% (by HPLC)
Catalog # ENZOBMLKI5900050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Bal.:
    50 µl
  • Sekvence:
    1202

Specifications

Specification Test Results

  • Formulation:
    Solution in DMSO
  • Absorbance maximum:
    350 - 380 nm
  • Emission maximum:
    450 - 480 nm
  • Purity:
    ≥95% (HPLC)

Specification Test Results

About this item

 :
 :GHS07
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)