ContactUsLinkComponent
Chromogen/Substrate Set
Chromogen/Substrate Set
Catalog # ENZOENZACC1050200
Supplier:  Enzo Life Sciences
CAS Number:  
Chromogen/Substrate Set
Catalog # ENZOENZACC1050200
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    200 mL
  • Protein/peptide name:
    Chromogen/substrate buffer
Specifications
About this item
The HIGHDEF® DAB Chromogen/Substrate Set can be used in conjunction with peroxidase-based immunostaining systems. Peroxidase from the antibody detection system reacts with hydrogen peroxide substrate to degrade it, which then reacts with DAB, precipitating it at positive sites and yielding a dark brown color.
  • Strong, clear color development
  • Compatible with high-throughput systems
3,3'-Diaminobenzidine (DAB), a widely used chromogen for immunoperoxidase staining, is well accepted among pathologists because of its increased sensitivity and decreased background when compared to amino ethyl carbazole (AEC). Specimens stained with DAB can be dehydrated, cleared, and mounted for permanent record keeping. HIGHDEF® DAB Chromogen/Substrate Set is more sensitive and stable than traditional working DAB solutions.