DiIC1(5) iodide ≥98% (by HPLC), Ultrapure
Catalog # ENZOENZ52207
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:510.5 g/mol
Storage Temperature:Freezer
CAS Number:36536-22-8
Formula:C₂₇H₃₁IN₂