Order Entry
Czech Republic
ContactUsLinkComponent
Benzamil hydrochloride ≥98% (by HPLC), yellow solid
Irritant
Benzamil hydrochloride ≥98% (by HPLC), yellow solid
Catalog # ENZOBMLNA1030050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
Benzamil hydrochloride ≥98% (by HPLC), yellow solid
Catalog # ENZOBMLNA1030050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :356.21 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:161804-20-2
 :2898-76-2
Formula:C₁₃H₁₄ClN₇O·HCl
MDL Number:MFCD00069219