Order Entry
Czech Republic
ContactUsLinkComponent
 
5-(N-Ethyl-N-isopropyl)amiloride ≥98% (by TLC)
Catalog # ENZOALX550266M005
Supplier:  Enzo Life Sciences
CAS Number:  
5-(N-Ethyl-N-isopropyl)amiloride ≥98% (by TLC)
Catalog # ENZOALX550266M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :299.763 g/mol
Storage Temperature:Refrigerator
CAS Number:1154-25-2
Formula:C₁₁H₁₈ClN₇O
MDL Number:MFCD00151710