N-(6-Aminohexyl)-5-chloro-1-naphthalenesulphonamide hydrochloride ≥98% (by TLC)
Catalog # ENZOBMLCA3200050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:377.32 g/mol
Storage Temperature:Freezer
CAS Number:61714-27-0
Formula:C₁₆H₂₁ClN₂O₂S
MDL Number:MFCD00012559