Order Entry
Czech Republic
ContactUsLinkComponent
Polygodial ≥98% (by HPLC)
Polygodial ≥98% (by HPLC)
Supplier:  Enzo Life Sciences
Certificates
Polygodial ≥98% (by HPLC)
Supplier:  Enzo Life Sciences

Specification Test Results

  • Source:
    Isolated from Polygonum hydropiper
  • Purity:
    ≥98% (HPLC)

Specification Test Results

Frequently Bought Together