Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gabapentin
Gabapentin ≥99% (by TLC)
Gabapentin ≥99% (by TLC)
Gabapentin ≥99% (by TLC)
Catalog # ENZOBMLCA2340100
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Gabapentin ≥99% (by TLC)
Catalog # ENZOBMLCA2340100
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
171.24 g/mol
CAS Number:
60142-96-3
Formula:
C₉H₁₇NO₂
MDL Number:
MFCD00865286
Product Details