SKF-96365 hydrochloride ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:130495-35-1
:
Formula:C₂₂H₂₆N₂O₃·HCl
MDL Number:MFCD00236407
:402.9 g/mol
Melting Pt:117…119 °C
Storage Temperature:Ambient
Frequently Bought Together