Order Entry
Czech Republic
ContactUsLinkComponent
Carvedilol ≥98% (by HPLC)
Environment
Carvedilol ≥98% (by HPLC)
Catalog # ENZOBMLAR1120100
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Environment
Carvedilol ≥98% (by HPLC)
Catalog # ENZOBMLAR1120100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :406.48 g/mol
Storage Temperature:Refrigerator
UN:3077
ADR:9,III
 :GHS09
CAS Number:72956-09-3
Formula:C₂₄H₂₆N₂O₄
MDL Number:MFCD00864692