Order Entry
Czech Republic
ContactUsLinkComponent
Ascomycin (from Streptomyces species) ≥90% (by HPLC, TLC)
Ascomycin (from Streptomyces species) ≥90% (by HPLC, TLC)
Catalog # ENZOBMLA1650001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ascomycin (from Streptomyces species) ≥90% (by HPLC, TLC)
Catalog # ENZOBMLA1650001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :790 g/mol
Storage Temperature:Freezer
CAS Number:104987-12-4
Formula:C₄₃H₆₉NO₁₂
MDL Number:MFCD06198665