Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
HT-2 Toxin
HT-2 Toxin ≥98% (by TLC)
HT-2 Toxin ≥98% (by TLC)
:
ENZO LIFE SCIENCES
HT-2 Toxin ≥98% (by TLC)
:
ENZO LIFE SCIENCES
:
26934-87-2
:
:
C₂₂H₃₂O₈
:
424.5 g/mol
:
Freezer
:
2811
:
6.1,I
:
GHS06
:
GHS07
Source:
Semisynthetic, derived from T2 toxin from Fusarium tricinctum
Purity:
≥98% (TLC)
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection