Order Entry
Czech Republic
ContactUsLinkComponent
JC-1 iodide salt ≥98% (by HPLC), Ultrapure
JC-1 iodide salt ≥98% (by HPLC), Ultrapure
Catalog # ENZOALX620053M005
Supplier:  Enzo Life Sciences
CAS Number:  
JC-1 iodide salt ≥98% (by HPLC), Ultrapure
Catalog # ENZOALX620053M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :652.23 g/mol
Storage Temperature:Freezer
CAS Number:47729-63-5
 :3520-43-2
Formula:C₂₅H₂₇Cl₄IN₄
MDL Number:MFCD00188475