Monophosphoryl lipid A 0.5 mg/l in water (synthetic) ready-to-use
Catalog # ENZOALX581205C100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Seq:1286
Specifications
Specification Test Results
- Source:Synthetic
- Concentration:0.5 mg/ml
- Formulation:Liquid. Sterile, ready-to-use solution in pyrogen free double distilled water.
Specification Test Results
About this item
:
:1763.47 g/mol
Storage Temperature:Refrigerator
CAS Number:1246298-63-4
Formula:C₉₆H₁₈₄N₃O₂₂P