Order Entry
Czech Republic
ContactUsLinkComponent
PNU-120596 ≥98% (by HPLC)
PNU-120596 ≥98% (by HPLC)
Catalog # ENZOALX550526M005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
PNU-120596 ≥98% (by HPLC)
Catalog # ENZOALX550526M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)
  • Identity:
    Determined by MS and NMR

Specification Test Results

About this item

 :
 :311.7 g/mol
Storage Temperature:Freezer
CAS Number:501925-31-1
Formula:C₁₃H₁₄ClN₃O₄
MDL Number:MFCD00095313