Dimethyl-W84 dibromide ≥98% (by HPLC)
Katalogové číslo ENZOALX550400M005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity (HPLC):≥98%
Specification Test Results
About this item
:
:736.6 g/mol
Storage Temperature:Refrigerator
CAS Number:402475-33-6
Formula:C₃₄H₄₈Br₂N₄O₄