Order Entry
Czech Republic
ContactUsLinkComponent
Praziquantel ≥98% (by HPLC)
Praziquantel ≥98% (by HPLC)
Catalog # ENZOALX550321G005
Supplier:  Enzo Life Sciences
CAS Number:  
Praziquantel ≥98% (by HPLC)
Catalog # ENZOALX550321G005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%

Specification Test Results

About this item

 :
 :312.41 g/mol
Merck Index:14,07716
Storage Temperature:Ambient
CAS Number:55268-74-1
Formula:C₁₉H₂₄N₂O₂
MDL Number:MFCD00058531