Order Entry
Czech Republic
ContactUsLinkComponent
Ruthenium(III) chloride oxide ≥90% (metals basis)
Ruthenium(III) chloride oxide ≥90% (metals basis)
Catalog # ENZOALX550295G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ruthenium(III) chloride oxide ≥90% (metals basis)
Catalog # ENZOALX550295G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :786.354 g/mol
Storage Temperature:Ambient
CAS Number:11103-72-3
Formula:H₄₂Cl₆N₁₄O₂Ru₃
MDL Number:MFCD00011479