Order Entry
Czech Republic
ContactUsLinkComponent
R-(+)-Baclofen hydrochloride ≥98% (by HPLC)
R-(+)-Baclofen hydrochloride ≥98% (by HPLC)
Katalogové číslo ENZOALX550074M050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
R-(+)-Baclofen hydrochloride ≥98% (by HPLC)
Katalogové číslo ENZOALX550074M050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%

Specification Test Results

About this item

 :
 :250.2 g/mol
Merck Index:14,00937
Storage Temperature:Refrigerator
CAS Number:63701-55-3
Formula:C₁₀H₁₂ClNO₂·HCl