0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Water
Water endotoxin-free, double distilled
Water endotoxin-free, double distilled
Catalog # ENZOALX505008LD15
Dodavatel:
ENZO LIFE SCIENCES
Číslo CAS:
Expand image
Water endotoxin-free, double distilled
Catalog # ENZOALX505008LD15
Dodavatel:
ENZO LIFE SCIENCES
Číslo CAS:
Specifikace
Pk:
1,5 mL
Seq:
2095
Specifikace
O tomto produktu
Product Details
:
:
18.02 g/mol
Číslo CAS:
7732-18-5
Vzorec:
H₂O
MDL číslo:
MFCD00011332
Product Details