5-Aza-2'-deoxycytidine ≥98% (by HPLC)
Catalog # ENZOALX480096M005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:228.21 g/mol
CAS Number:2353-33-5
Formula:C₈H₁₂N₄O₄
MDL Number:MFCD00043011