8-(4-Chlorophenylthio)-2'-O-methyladenosine-3,5-cyclic monophosphate sodium salt ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:510774-50-2
:
Formula:C₁₇H₁₆ClN₅O₆PS·Na
:507.8 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity (HPLC):≥98%
- Identity:Determined by ESI-MS and UV
- Purity detail:Product is not sterile
Specification Test Results
Frequently Bought Together