Order Entry
Czech Republic
ContactUsLinkComponent
8-Hydroxyguanosine ≥98% (by HPLC)
8-Hydroxyguanosine ≥98% (by HPLC)
Catalog # ENZOALX480075M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
8-Hydroxyguanosine ≥98% (by HPLC)
Catalog # ENZOALX480075M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%
  • Identity:
    Determined by UV-VIS and ESI-MS

Specification Test Results

About this item

 :
 :299.243 g/mol
Storage Temperature:Freezer
CAS Number:3868-31-3
Formula:C₁₀H₁₃N₅O₆