Guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥98% (by HPLC)
Catalog # ENZOALX480072M001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:360.3 · 23.0 g/mol
Storage Temperature:Refrigerator
CAS Number:86562-09-6
Formula:C₁₀H₁₁N₅O₆SP·Na