Order Entry
Czech Republic
ContactUsLinkComponent
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥90% (by HPLC) K+ channel activator
Irritant
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥90% (by HPLC) K+ channel activator
Catalog # ENZOALX480066M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Irritant
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥90% (by HPLC) K+ channel activator
Catalog # ENZOALX480066M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :546.98 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:93839-89-5
Formula:C₁₀H₁₂N₅O₁₂PS·4Li
MDL Number:MFCD00077293