Order Entry
Czech Republic
ContactUsLinkComponent
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate triethylammonium salt ≥99% (by HPLC)
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate triethylammonium salt ≥99% (by HPLC)
Catalog # ENZOALX480031M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate triethylammonium salt ≥99% (by HPLC)
Catalog # ENZOALX480031M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥99%
  • Identity:
    Determined by MS and UV

Specification Test Results

About this item

 :
 :589 g/mol
Storage Temperature:Freezer
CAS Number:54364-02-2
Formula:C₁₆H₁₄ClN₅O₇PS·C₆H₁₆N