
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine ≥97% (by non aqueous titration)
Catalog # ENZOALX450011M100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Purity (Nonaqueous titration):≥97%
- Appearance:White to off-white solid
Specification Test Results
About this item
:
:424.55 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:16858-02-9
Formula:C₂₆H₂₈N₆
MDL Number:MFCD00036918