

DEA NONOate ≥98% (by NMR), white to off-white solid
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:372965-00-9
:
Formula:C₄H₁₀N₃O₂·C₄H₁₂N
:206.2 g/mol
:Packaged under inert gas.
:GHS05
:GHS07
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
Specification Test Results
- Purity (NMR):≥98%
Specification Test Results
Frequently Bought Together