Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Noc-12
Noc-12 ≥97% (by HPLC)
Noc-12 ≥97% (by HPLC)
:
ENZO LIFE SCIENCES
Noc-12 ≥97% (by HPLC)
:
ENZO LIFE SCIENCES
:
146724-89-2
:
:
C₆H₁₆N₄O₂
:
176.2 g/mol
:
Packaged under inert gas.
:
GHS07
:
Freezer
Purity:
≥97% (HPLC)
Frequently Bought Together
VWR®, Water Sampling Bottles, Sterile
: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
: VWR Collection
VWR® Blender Bags
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
Petri dishes, Corning® Gosselin™
: Corning
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
Storage Boxes
: EPPENDORF
VWR®, Ice Pans, with or without Lid
: VWR Collection
Deepwell Plates 96-Well, 2000 µl
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
VWR® Bottles, Water Sampling
: VWR Collection