
DETA NONOate (NOC-18) ≥97% (by 1H-NMR)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:146724-94-9
:
Formula:C₄H₁₃N₅O₂
MDL Number:MFCD00278808
:163.181 g/mol
:GHS07
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
Specification Test Results
- Purity:≥97% (1H-NMR)
Specification Test Results
Frequently Bought Together