GEA 5583 ≥97% (by HPLC)
Catalog # ENZOALX430006M010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Purity:≥97% (HPLC)
- Identity:Determined by IR
Specification Test Results
About this item
:
:292.7 g/mol
Storage Temperature:Refrigerator
Formula:C₁₁H₉ClN₆O₂