

Sodium diethyldithiocarbamate trihydrate ≥99% (by titrimetric analysis)
Catalog # ENZOALX400003G025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:225.31 g/mol
Merck Index:12,03443
Melting Pt:95…98.5 °C
Density:1,1 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:205-710-6
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:20624-25-3
Formula:(C₂H₅)₂NCSSNa·3H₂O
MDL Number:MFCD00150617