Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium 1-pyrrolidinecarbodithioate ≥98% (by HPLC)
Ammonium 1-pyrrolidinecarbodithioate ≥98% (by HPLC)
Catalog # ENZOALX400002G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ammonium 1-pyrrolidinecarbodithioate ≥98% (by HPLC)
Catalog # ENZOALX400002G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :146.3 · 18.0 g/mol
Storage Temperature:Refrigerator
EINECS:225-834-4
CAS Number:5108-96-3
Formula:C₅H₁₂N₂S₂
MDL Number:MFCD00012720