Zadání objednávky
Czech Republic
ContactUsLinkComponent
Wogonin ≥98% (by HPLC)
Corrosive
Wogonin ≥98% (by HPLC)
Catalog # ENZOALX385033M025
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Corrosive
Wogonin ≥98% (by HPLC)
Catalog # ENZOALX385033M025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Bal.:
    25 mg
  • Sekvence:
    2315

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)
  • Identity:
    Determined by ¹H-NMR

Specification Test Results

About this item

 :
 :284.3 g/mol
Storage Temperature:Refrigerator
UN:2790
ADR:8,III
 :GHS05
CAS Number:632-85-9
Formula:C₁₆H₁₂O₅
MDL Number:MFCD00017736