Order Entry
Czech Republic
ContactUsLinkComponent
Quercetin dihydrate ≥95% (by HPLC)
Quercetin dihydrate ≥95% (by HPLC)
Catalog # ENZOALX385001G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Quercetin dihydrate ≥95% (by HPLC)
Catalog # ENZOALX385001G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :338.268 g/mol
CAS Number:6151-25-3
Formula:C₁₅H₁₄O₉
MDL Number:MFCD00149487