Order Entry
Czech Republic
ContactUsLinkComponent
Enniatin A (from Fusarium sp.) ≥97% (by HPLC)
Enniatin A (from Fusarium sp.) ≥97% (by HPLC)
Catalog # ENZOALX380310MC05
Supplier:  Enzo Life Sciences
CAS Number:  
Enniatin A (from Fusarium sp.) ≥97% (by HPLC)
Catalog # ENZOALX380310MC05
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Seq:
    2035

Specifications

About this item

 :
 :681.9 g/mol
Storage Temperature:Freezer
CAS Number:2503-13-1
Formula:C₃₆H₆₃N₃O₉