Order Entry
Czech Republic
ContactUsLinkComponent
 
Sparfloxacin ≥98%
Catalog # ENZOALX380302G005
Supplier:  Enzo Life Sciences
CAS Number:  
Sparfloxacin ≥98%
Catalog # ENZOALX380302G005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :392.407 g/mol
CAS Number:110871-86-8
Formula:C₁₉H₂₂F₂N₄O₃
MDL Number:MFCD00869619